Merck
  • Targeting the integrated networks of aggresome formation, proteasome, and autophagy potentiates ER stress‑mediated cell death in multiple myeloma cells.
Targeting the integrated networks of aggresome formation, proteasome, and autophagy potentiates ER stress‑mediated cell death in multiple myeloma cells.
International journal of oncology (2014-11-26)
Shota Moriya, Seiichiro Komatsu, Kaho Yamasaki, Yusuke Kawai, Hiroko Kokuba, Ayako Hirota, Xiao-Fang Che, Masato Inazu, Akihiko Gotoh, Masaki Hiramoto, Keisuke Miyazawa
ABSTRACT
The inhibitory effects of macrolide antibiotics including clarithromycin (CAM) on autophagy flux have been reported. Although a macrolide antibiotic exhibits no cytotoxicity, its combination with bortezomib (BZ), a proteasome inhibitor, for the simultaneous blocking of the ubiquitin (Ub)‑proteasome and autophagy‑lysosome pathways leads to enhanced multiple myeloma (MM) cell apoptosis induction via stress overloading of the endoplasmic reticulum (ER). As misfolded protein cargo is recruited by histone deacetylase 6 (HDAC6) to dynein motors for aggresome transport, serving to sequester misfolded proteins, we further investigated the cellular effects of targeting proteolytic pathways and aggresome formation concomitantly in MM cells. Pronounced apoptosis was induced by the combination of vorinostat [suberoylanilide hydroxamic acid (SAHA); potently inhibits HDAC6] with CAM and BZ compared with each reagent or a 2‑reagent combination. CAM/BZ treatment induced vimentin positive‑aggresome formation along with the accumulation of autolysosomes in the perinuclear region, whereas they were inhibited in the presence of SAHA. The SAHA/CAM/BZ combination treatment maximally upregulated genes related to ER stress including C/EBP homologous protein (CHOP). Similarly to MM cell lines, enhanced cytotoxicity with CHOP upregulation following SAHA/CAM/BZ treatment was shown by a wild‑type murine embryonic fibroblast (MEF) cell line; however, a CHOP‑deficient MEF cell line almost completely canceled this pronounced cytotoxicity. Knockdown of HDAC6 with siRNA exhibited further enhanced CAM/BZ‑induced cytotoxicity and CHOP induction along with the cancellation of aggresome formation. Targeting the integrated networks of aggresome, proteasome, and autophagy is suggested to induce efficient ER stress‑mediated apoptosis in MM cells.
Product Description
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ first grade, ≥95.0%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Supelco
Methanol, analytical standard
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Methanol, SAJ special grade
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Methanol, suitable for HPLC
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPriceQuantity
OR14608-1G1 g
Ships in 14 days - from Aldrich Partner.
CHF 98.30
OR14608-5G5 g
Ships in 14 days - from Aldrich Partner.
CHF 273.20