Merck
  • New microfluidic-based sampling procedure for overcoming the hematocrit problem associated with dried blood spot analysis.
New microfluidic-based sampling procedure for overcoming the hematocrit problem associated with dried blood spot analysis.
Analytical chemistry (2015-01-22)
Luc Alexis Leuthold, Olivier Heudi, Julien Déglon, Marc Raccuglia, Marc Augsburger, Franck Picard, Olivier Kretz, Aurélien Thomas
ABSTRACT
Hematocrit (Hct) is one of the most critical issues associated with the bioanalytical methods used for dried blood spot (DBS) sample analysis. Because Hct determines the viscosity of blood, it may affect the spreading of blood onto the filter paper. Hence, accurate quantitative data can only be obtained if the size of the paper filter extracted contains a fixed blood volume. We describe for the first time a microfluidic-based sampling procedure to enable accurate blood volume collection on commercially available DBS cards. The system allows the collection of a controlled volume of blood (e.g., 5 or 10 μL) within several seconds. Reproducibility of the sampling volume was examined in vivo on capillary blood by quantifying caffeine and paraxanthine on 5 different extracted DBS spots at two different time points and in vitro with a test compound, Mavoglurant, on 10 different spots at two Hct levels. Entire spots were extracted. In addition, the accuracy and precision (n = 3) data for the Mavoglurant quantitation in blood with Hct levels between 26% and 62% were evaluated. The interspot precision data were below 9.0%, which was equivalent to that of a manually spotted volume with a pipet. No Hct effect was observed in the quantitative results obtained for Hct levels from 26% to 62%. These data indicate that our microfluidic-based sampling procedure is accurate and precise and that the analysis of Mavoglurant is not affected by the Hct values. This provides a simple procedure for DBS sampling with a fixed volume of capillary blood, which could eliminate the recurrent Hct issue linked to DBS sample analysis.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
SKU
Pack Size
Availability
Price
Quantity
Supelco
Methanol, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ special grade
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, NMR reference standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, Absolute - Acetone free
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%