Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Pricing and availability is not currently available.
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Pricing and availability is not currently available.
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Glycine, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Pricing and availability is not currently available.
Supelco
Methanol, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, SAJ special grade
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Glycine, 99%, FCC
Pricing and availability is not currently available.
Pricing and availability is not currently available.
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Pricing and availability is not currently available.
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Pricing and availability is not currently available.
Sigma-Aldrich
Glycine, SAJ special grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Methanol, NMR reference standard
Pricing and availability is not currently available.