Merck
  • Ostruthin: an antimycobacterial coumarin from the roots of Peucedanum ostruthium.
Ostruthin: an antimycobacterial coumarin from the roots of Peucedanum ostruthium.
Planta medica (2003-04-24)
Andreas Schinkovitz, Simon Gibbons, Michael Stavri, Michael J Cocksedge, Franz Bucar
ABSTRACT
Following a bioassay-guided fractionation, ostruthin (6-geranyl-7-hydroxycoumarin) was isolated from the roots of Peucedanum ostruthium Koch (Apiaceae) as a compound with pronounced in vitro activity against several species of rapidly growing Mycobacteria, namely Mycobacterium abscesus, M. aurum, M. fortuitum, M. phlei and M. smegmatis. Minimum inhibitory concentrations (MIC) ranged between 3.4 to 107.4 microM and were comparable to those of ethambutol and isoniazid. Imperatorin (8-isopent-2-enyloxy-6,7-furanocoumarin) showed no activity at concentrations up to 1.9 mM. Umbelliferone (7-hydroxycoumarin) was only weakly active (MIC = 0.79 mM).
Sigma-Aldrich
6-Geranyl-7-hydroxycoumarin, ≥95% (LC/MS-ELSD)
Pricing and availability is not currently available.