- Tritrichomonas foetus infection in cats with diarrhoea in a rescue colony in Italy.
Tritrichomonas foetus infection in cats with diarrhoea in a rescue colony in Italy.
Journal of feline medicine and surgery (2008-09-09)
Malcolm Holliday, Dario Deni, Danièlle A Gunn-Moore
PMID18774326
ABSTRACT
Faecal samples from 74 cats with chronic large bowel diarrhoea that were living in a rescue colony in Italy, were submitted for assessment of Tritrichomonas foetus by direct microscopy of faecal smears (n=20) and T foetus-specific culture (n=74), with confirmation by polymerase chain reaction (n=1). Twenty-four of the cats (32%) were found to be infected with T foetus. The infected cats were predominantly over a year of age (67%) and were all neutered non-pedigree domestic cats.
MATERIALS