Merck
Synthesis of sporiolide B from D-glucal.
Carbohydrate research (2007-05-23)
Qi Chen, Yuguo Du
ABSTRACT
A total synthesis of the 12-membered ring natural macrolide, sporiolide B, was achieved from D-glucal in 17 steps with 4.8% overall yield. The required stereochemical configuration at C-3 and C-5 in sporiolide B was easily introduced by applying a Mitsunobu reaction on the chiral template D-glucal. Yamaguchi esterification and ring closing metathesis greatly improved the access to the target compound.
Sigma-Aldrich
D-Glucal, 96%
Pricing and availability is not currently available.