Merck
Production responses to internal parasite control in dairy cattle.
Australian veterinary journal (1992-09-01)
S A Spence, G C Fraser, E B Dettmann, D F Battese
ABSTRACT
A significant increase in milk production, averaging 164 litres per cow per lactation (a 4.8% increase), was seen after cows infected with gastrointestinal nematodes, paramphistomes and Fasciola hepatica were treated with broad-spectrum anthelmintics. Three hundred and ninety pairs of cows from eight herds with year-round calving were studied. One cow in each matched pair was given 7.5 mg/kg fenbendazole, 7.5 mg/kg levamisole hydrochloride and 15 mg/kg oxyclozanide in March, May and August of one year; the other cow in the pair received no anthelmintic. The number of nematode and trematode eggs was significantly decreased in the faeces of treated cows.
Product Description
Supelco
Oxyclozanide, VETRANAL®, analytical standard
Pricing and availability is not currently available.