Merck
  • Protocol to generate and utilize pancreatic tissue slices to study endocrine and exocrine physiology in situ from mouse and human tissue.
Protocol to generate and utilize pancreatic tissue slices to study endocrine and exocrine physiology in situ from mouse and human tissue.
STAR protocols (2023-07-01)
Julia K Panzer, Alejandro Caicedo
ABSTRACT
Pancreatic tissue slices allow functional investigations under close physiological conditions in situ. This approach is particularly advantageous for studying infiltrated and structurally damaged islets as found in T1D. More importantly, slices allow studying the interplay between endocrine and exocrine compartments. We here describe how to perform agarose injections, tissue preparation, and slice procedure for mouse and human tissue. We then describe in detail how to use the slices to perform functional studies using hormone secretion and calcium imaging as readouts. For complete details on the use and execution of this protocol, please refer to Panzer et al. (2022).1.
Sigma-Aldrich
(−)-Epinephrine (+)-bitartrate salt, solid
Sigma-Aldrich
Monoclonal Anti-Glucagon antibody produced in mouse, clone K79bB10, ascites fluid
Sigma-Aldrich
Anti-Somatostatin Antibody, clone YC7, culture supernatant, clone YC7, Chemicon®