Merck
  • An Orthogonal Approach for Determination of Acetamide Content in Pharmaceutical Drug Substance and Base-Contaminated Acetonitrile by GC and GC-MS External Method.
An Orthogonal Approach for Determination of Acetamide Content in Pharmaceutical Drug Substance and Base-Contaminated Acetonitrile by GC and GC-MS External Method.
Journal of chromatographic science (2019-09-11)
Nagaraju Rajana, Kaviaraj M Yarbagi, K Balakumaran, M V Madhubabu, J Mosesbabu, K Basavaiah, Dharamasoth Rama Devi
ABSTRACT
Acetamide is a potential genotoxic impurity; it should control in drug substance based on daily dosage level. It forms from base-contaminated acetonitrile and by-product of some drug substances. The available methods for acetamide in drug substance and water samples were determined by GC-MS using internal standard with critical procedures. These developed and validated methods can assist in evaluating the reaction between acetonitrile and different bases and also determine trace level acetamide in drug substances. The method development was initiated with DB-624, 30 m, 0.32 width and 1.0-μm column. The column was used to validate at the 600 ppm TTC value. Similarly, the CP-SIL 5CB, 60 m, 0.32 width, the 5-μm column was used for the remaining TTC values. The validation study was performed for all TTC limits. The % RSD for precision at 600, 60, 20, 10 and 2.5 ppm was <15%. The % recovery at all TTC level was in between the 70 and 130%. Solution stability study was performed up to the 24 h. At 2.5 ppm, the results were <15% variation from the initial value. The linearities from the 50 to 150% concerning TTC values were more than limit of 0.98 correlation coefficient. The limit of detection and limit of quantitation values were 0.4 to the 1.3 ppm, respectively, for 2.5 ppm TTC limit method.
Sigma-Aldrich
2-Methylpyrrolidine, 96%
Pricing and availability is not currently available.
Sigma-Aldrich
Pyrimidine, ≥98.0%
Pricing and availability is not currently available.