Merck
  • Adenosine A2B and A3 receptor location at the mouse neuromuscular junction.
Adenosine A2B and A3 receptor location at the mouse neuromuscular junction.
Journal of anatomy (2014-04-24)
Neus Garcia, Mercedes Priego, Erica Hurtado, Teresa Obis, Manel M Santafe, Marta Tomàs, Maria Angel Lanuza, Josep Tomàs
ABSTRACT
To date, four subtypes of adenosine receptors have been cloned (A(1)R, A(2A)R, A(2B)R, and A(3)R). In a previous study we used confocal immunocytochemistry to identify A(1)R and A(2A)R receptors at mouse neuromuscular junctions (NMJs). The data shows that these receptors are localized differently in the three cells (muscle, nerve and glia) that configure the NMJs. A(1)R localizes in the terminal teloglial Schwann cell and nerve terminal, whereas A(2A)R localizes in the postsynaptic muscle and in the axon and nerve terminal. Here, we use Western blotting to investigate the presence of A(2B)R and A(3)R receptors in striated muscle and immunohistochemistry to localize them in the three cells of the adult neuromuscular synapse. The data show that A(2B)R and A(3)R receptors are present in the nerve terminal and muscle cells at the NMJs. Neither A(2B)R nor A(3)R receptors are localized in the Schwann cells. Thus, the four subtypes of adenosine receptors are present in the motor endings. The presence of these receptors in the neuromuscular synapse allows the receptors to be involved in the modulation of transmitter release.
Sigma-Aldrich
Anti-Adenosine A3 Receptor Antibody, extracellular (a.a. 154-168), Chemicon®, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Adenosine A2b Receptor Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.