Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

V900722

Sigma-Aldrich

4-Formylbenzoic acid

Vetec, reagent grade, 97%

Synonym(s):

4-Carboxybenzaldehyde, Benzaldehyde-4-carboxylic acid, Terephthalaldehydic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CC6H4CHO
CAS Number:
Molecular Weight:
150.13
Beilstein:
471734
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

97%

mp

247 °C (lit.)

SMILES string

[H]C(=O)c1ccc(cc1)C(O)=O

InChI

1S/C8H6O3/c9-5-6-1-3-7(4-2-6)8(10)11/h1-5H,(H,10,11)

InChI key

GOUHYARYYWKXHS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

V900722-BULK:
V900722-VAR:
V900722-25G:
V900722-100G:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yves Meyer et al.
Organic & biomolecular chemistry, 8(8), 1777-1780 (2010-05-08)
This study focuses on the disassembly-behavior of self-immolative pro-fluorescent linkers under physiological conditions and through an enzyme-initiated domino reaction. The targeted linkers are based on para-aminobenzylalcohol (PABA) or hemithioaminal derivatives of para-carboxybenzaldehyde or glyoxilic acid. We found that a fine
A Magnien et al.
European journal of biochemistry, 131(2), 375-381 (1983-03-15)
Initial-rate measurements were made of the reduction of pyridine-3-aldehyde and p-carboxybenzaldehyde by NADPH catalyzed by pig liver aldehyde reductase I. The initial velocity analysis and product inhibition data suggest that aldehyde reductase I obeys a compulsory-order mechanism with pyridine-3-aldehyde as
C M Ryle et al.
The Biochemical journal, 197(3), 715-720 (1981-09-01)
The distribution of the two principal isoenzymes of aldehyde reductase (EC 1.1.1.2) has been studied in ox brain. The more active of these, which has been termed the high-Km enzyme, has been shown to be located in the cytosol and
Xiaofei Chen et al.
Biomaterials science, 3(6), 870-878 (2015-07-30)
Herein, hyperbranched poly(ethylene glycol)-based supramolecular nanoparticles with pH-sensitive properties were designed and used for targeted drug delivery. Via host-guest recognition between benzimidazole anchored poly(ethylene glycol)-hyperbranched polyglycerol (PEG-HPG-BM) and folic acid modified CD (FA-CD), targeted supramolecular nanoparticles (TSNs) were fabricated. At
Da Huang et al.
Biomaterials, 52, 417-425 (2015-03-31)
Bone tuberculosis (TB) is one of the most common extrapulmonary TB. Effective integration of chemotherapy and bone regeneration is an optimal solution for bone TB therapy. Herein, we produce a composite scaffold drug delivery system fabricated with an isoniazid conjugated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service