Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

V800264

Sigma-Aldrich

Molybdic acid

LR, ≥85%

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Product V800264 is not currently sold in your country. Contact Technical Service

grade

LR

vapor density

6.2 (vs air)

product line

Vetec

Assay

≥85%

form

powder

solubility

water: insoluble

density

3.1 g/mL at 25 °C (lit.)

SMILES string

O[Mo](O)(=O)=O

InChI

1S/Mo.2H2O.2O/h;2*1H2;;/q+2;;;;/p-2

InChI key

VLAPMBHFAWRUQP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

V800264-VAR:
V800264-500G:4548173325118
V800264-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service