Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

V800257

Sigma-Aldrich

Mercury

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Hg
CAS Number:
Molecular Weight:
200.59
EC Number:
MDL number:
UNSPSC Code:
12141726
PubChem Substance ID:
Assay:
≥99%
form:
liquid
Product V800257 is not currently sold in your country. Contact Technical Service

vapor density

7 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)
1 mmHg ( 126 °C)

product line

Vetec

Assay

≥99%

form

liquid

resistivity

95.8 μΩ-cm, 20°C

bp

356.6 °C (lit.)

mp

−38.87 °C (lit.)

cation traces

Fe: ≤0.0001%
Pb: ≤0.0003%

SMILES string

[Hg]

InChI

1S/Hg

InChI key

QSHDDOUJBYECFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

V800257-500G:4548173325040
V800257-BULK:
V800257-VAR:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabrina Llop et al.
PloS one, 9(5), e97172-e97172 (2014-05-17)
The genetic background may influence methylmercury (MeHg) metabolism and neurotoxicity. ATP binding cassette (ABC) transporters actively transport various xenobiotics across biological membranes. To investigate the role of ABC polymorphisms as modifiers of prenatal exposure to MeHg. The study population consisted
Birgit M Braune et al.
Environment international, 66, 92-96 (2014-02-22)
Mercury (Hg) is a potentially toxic metal ubiquitous in arctic biota. Livers of adult thick-billed murres (Uria lomvia) and northern fulmars (Fulmarus glacialis) sampled from several locations in the eastern Canadian Arctic during 2007-2008 were analyzed for total Hg in
Nadine Czekalski et al.
The ISME journal, 8(7), 1381-1390 (2014-03-07)
Antibiotic-resistance genes (ARGs) are currently discussed as emerging environmental contaminants. Hospital and municipal sewage are important sources of ARGs for the receiving freshwater bodies. We investigated the spatial distribution of different ARGs (sul1, sul2, tet(B), tet(M), tet(W) and qnrA) in
James P K Rooney
Toxicology and applied pharmacology, 274(3), 425-435 (2013-12-26)
Reports from human case studies indicate a half-life for inorganic mercury in the brain in the order of years-contradicting older radioisotope studies that estimated half-lives in the order of weeks to months in duration. This study systematically reviews available evidence
Okechukwu S Ogah et al.
JACC. Heart failure, 2(3), 250-259 (2014-06-24)
The aim of this study was to determine the contemporary profile, clinical characteristics, and intrahospital outcomes of acute heart failure (AHF) in an African urban community. There are limited data on the current burden and characteristics of AHF in Nigerian

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service