Skip to Content
Merck
All Photos(1)

Documents

Safety Information

V800106

Sigma-Aldrich

Calcium hypochlorite

LR, available chlorine ≥30 %

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(OCl)2
CAS Number:
Molecular Weight:
142.98
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

LR

product line

Vetec

form

powder

composition

available chlorine, ≥30%

mp

100 °C (lit.)

density

2.35 g/mL at 25 °C (lit.)

SMILES string

ClO[Ca]OCl

InChI

1S/Ca.2ClO/c;2*1-2/q+2;2*-1

InChI key

ZKQDCIXGCQPQNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 1: Oxidizing solids
Hypochlorites
Hazardous rank I
1st oxidizing solid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

V800106-BULK:
V800106-VAR:
V800106-500G:4548173323220


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Boukhoubza et al.
Journal of environmental management, 91(1), 124-132 (2009-09-02)
The application of hypochlorite for the removal of soluble COD, phenolic and polyphenolic like compounds, and other organic compounds responsible for the olive mill wastewater (OOWW) colour has been experimentally studied. After the OOWW filtration on a sand column, the
Peter K Jensen et al.
Journal of health, population, and nutrition, 21(1), 26-31 (2003-05-20)
To evaluate the importance of public-domain transmission of pathogens in drinking-water, an intervention study was carried out by chlorinating the public water-supply system in a village in Pakistan. The water quality improved and reached a geometric mean of 3 Escherichia
M L Bari et al.
Journal of food protection, 73(4), 752-757 (2010-04-10)
The majority of the seed sprout-related outbreaks have been associated with Escherichia coli O157:H7 and Salmonella. Therefore, an effective method for inactivating these organisms on the seeds before sprouting is needed. The current pasteurization method for mung beans in Japan
Jed W Fahey et al.
Nutrition journal, 5, 13-13 (2006-04-25)
The recent increased interest in consuming green vegetable sprouts has been tempered by the fact that fresh sprouts can in some cases be vehicles for food-borne illnesses. They must be grown according to proper conditions of sanitation and handled as
Hudaa Neetoo et al.
International journal of food microbiology, 131(2-3), 218-223 (2009-04-03)
The application of high hydrostatic pressure technology as a seed decontamination technology was evaluated. Alfalfa seeds inoculated with approximately 10(5) CFU/g of Escherichia coli O157:H7 were subjected to oscillatory pressure treatments at 600 MPa and 20 degrees C for up

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service