Skip to Content
Merck
All Photos(1)

Key Documents

TA9H98771495

Targetmol Chemicals Inc.

TRAM 39

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
303.79

Iupac Name

2-(2-chlorophenyl)-2,2-diphenylacetonitrile

Inchi Code

1S/C20H14ClN/c21-19-14-8-7-13-18(19)20(15-22,16-9-3-1-4-10-16)17-11-5-2-6-12-17/h1-14H

InChI key

JHNRTPKGSCVKKC-UHFFFAOYSA-N

storage temp.

-80/-20

Purity

99.51%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/TRAM 39

MSDS

https://www.targetmol.com/compound/TRAM 39


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library