Safety Information
SU860106
Supelco
Crimp seals with Viton® septa
silver seal, magnetic (with 8mm center hole), diam. × thickness 20 mm × 1.0 mm, black Viton® septum, pkg of 100 ea
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
material
black Viton® septum
silver seal (magnetic with 8 mm center hole)
feature
closure type crimp seal
closure type crimp top vial
hardness
70 (Durometer: shore A)
packaging
pkg of 100 ea
diam. × thickness
20 mm × 1.0 mm
fitting
closure for crimp top vial
compatibility
for use with SPME
Legal Information
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)