Skip to Content
Merck
All Photos(1)

Documents

Safety Information

PHR9192

Supelco

Metoprolol EP Impurity N

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

3-[(1-Methylethyl)amino]-1,2-propanediol, 3-Isopropylamino-1,2-propanediol, 3-[(1-Methylethyl)amino]propane-1,2-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15NO2
CAS Number:
Molecular Weight:
133.19
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

Agency

Not traceable to any primary pharmacopoeia standard

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

application(s)

pharmaceutical small molecule

storage temp.

−20°C

SMILES string

OCC(O)CNC(C)C

InChI

1S/C6H15NO2/c1-5(2)7-3-6(9)4-8/h5-9H,3-4H2,1-2H3

InChI key

YKBZGEJJKPNRSI-UHFFFAOYSA-N

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Application

These Reference Materials are suitable for use in several analytical applications including, but not limited to, method development for qualitative and quantitative analyses, daily calibration, and routine quality control testing.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

PHR9192-50MG:
PHR9192-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service