Skip to Content
Merck
All Photos(1)

Key Documents

N12817

Paraquat CL tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107

grade

analytical standard

packaging

ampule of 500 membranes

manufacturer/tradename

Chem Service, Inc. N-12817-500MG

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sunitha Rangaraju et al.
eLife, 4, e08833-e08833 (2015-12-02)
Longevity mechanisms increase lifespan by counteracting the effects of aging. However, whether longevity mechanisms counteract the effects of aging continually throughout life, or whether they act during specific periods of life, preventing changes that precede mortality is unclear. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service