Skip to Content
Merck
All Photos(1)

Documents

G005715

HPLC Analysis of Organic Acids on Ascentis® RP-Amide

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Ascentis RP-Amide, 10 cm x 2.1 mm I.D., 3 μm particles (565301-U)
mobile phase: [A] 0.1% (v/v) TFA in [B] water
flow rate: 0.3 mL/min
column temp.: 35 °C
detector: UV, 210 nm
injection: 1 μL
sample: 0.2 - 4 g/L in mobile phase

suitability

application for HPLC

application(s)

chemicals and industrial polymers
food and beverages
vitamins, nutraceuticals, and natural products

Analysis Note

This application demonstrates the suitability of Ascentis RP-Amide for the analysis of organic acids under 100% aqueous conditions.

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Citric acid
    Acrylic acid
    Fumaric acid

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service