Skip to Content
Merck
All Photos(1)

Documents

Safety Information

98835

Supelco

4-Amino-3-methylphenol

analytical standard

Synonym(s):

2-Amino-5-hydroxytoluene, 4-Amino-m-cresol, 4-Hydroxy-2-methylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(CH3)OH
CAS Number:
Molecular Weight:
123.15
Beilstein:
2078803
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

176-179 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

Cc1cc(O)ccc1N

InChI

1S/C7H9NO/c1-5-4-6(9)2-3-7(5)8/h2-4,9H,8H2,1H3

InChI key

QGNGOGOOPUYKMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

98835-100MG:
98835-VAR:
98835-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service