Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

807455

TOYOPEARL® HW-50S Bulk Media

matrix hydroxylated methacrylate, bottle of 250 mL, 20-40 μm

Synonym(s):

TOYOPEARL® HW-50 Size Exclusion Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161501
Product 807455 is not currently sold in your country. Contact Technical Service

description

HW-50S

product line

TOYOPEARL®

form

slurry

packaging

bottle of 250 mL

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylate

matrix active group

polymer phase

particle size

20-40 μm

pore size

500-20,000 Da MW range

operating pH

2-13

separation technique

size exclusion (SEC)

General description

Toyopearl HW-50 resin is a hydroxylated methacrylic polymer for size exclusion chromatography of proteins between 500-80,000 Da.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

807455:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service