Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

69334

Supelco

(2,2-Dimethyl-3-methylsulfonyloxypropyl) methanesulfonate

analytical standard

Synonym(s):

Neopentyl glycol dimethylsulfate, 2,2-Dimethyl-1,3-propanediol dimethylsulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16O6S2
CAS Number:
Molecular Weight:
260.33
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

69-74 °C

format

neat

storage temp.

2-30°C

SMILES string

CC(C)(COS(C)(=O)=O)COS(C)(=O)=O

InChI

1S/C7H16O6S2/c1-7(2,5-12-14(3,8)9)6-13-15(4,10)11/h5-6H2,1-4H3

InChI key

MBBDNGMNEVIICT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

69334-VAR:
69334-100MG:
69334-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service