Skip to Content
Merck
All Photos(1)

Documents

61878-U

Proteomix® HIC Phenyl-NP5 (5 µm) HPLC Columns

L × I.D. 50 mm × 7.8 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

product name

Proteomix® HIC Phenyl-NP5, 5 μm, NP, L × I.D. 50 mm × 7.8 mm

material

stainless steel hardware

product line

Proteomix®

manufacturer/tradename

Sepax 433NP5-7805

packaging

1 ea of

parameter

6000 psi max. pressure
80 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

50 mm × 7.8 mm

impurities

<10 ppm metals

matrix

polymeric
polystyrene/divinylbenzene particle platform (Spherical, highly-crossed linked)

matrix active group

phenyl phase

particle size

5 μm

pore size

non-porous

operating pH range

2-12

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

Compatibility

Compatible with aqueous solution, a mixture of water and acetonitrile, acetone, methanol, or THF.

Analysis Note

Proteomix® HIC columns are specially designed for high resolution and high efficiency separations of proteins, oligonucleotides and peptides. Utilizing proprietary surface technologies, Proteomix HIC-NP resin is made of non-porous polystyrenedivinylbenzene (PS/DVB) beads with narrow-dispersed particle size distribution. The PS/DVB bead is modified with alkyl groups or aryl group that provides hydrophobic interaction with analytes. Proteomix HIC-NP resin is highly rigid and mechanically stable. In comparison to silica based HIC phase media, Proteomix HIC-NP phases have advantages for biomolecule separations with wide pH range (2-12) and high thermal stability. The nonporous structure and narrow particle distribution offer special selectivity, high resolution separation of proteins such as mAb (monoclonal antibody), ADC (antibody drug conjugate) and related protein fragments, DNA and oligonucleotides. Proteomix HIC-NP media is applicable at laboratory discovery, laboratory-scale purification and process chromatography for the production of a few mgs to kilogram of proteins.

Other Notes

Related catagories: Analytical/Chromatography, HPLC, HPLC & FPLC Columns for Biopolymer Separations, Sepax Technologies UHPLC Columns, Zenix Gel Filtration Columns (3 micron), Zenix-C SEC-100 Gel Filtration Column, 3 Micron, Zenix-C SEC-100 Guard Column, 3 micron

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Proteomix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service