Safety Information
58462
Supelco
Dynaseal Fittings
Set of nuts, collet, and ferrule
About This Item
UNSPSC Code:
41115700
packaging
pkg of 2 ea
fitting
10-32 male Dynaseal nut and ferrule for 1/16 in. O.D. tubing (collet included)
Features and Benefits
Dynaseal hand-tight fittings will seal against pressures to over 7000psi/490kg/cm². Each fitting consists of a reusable nut, collet, and ferrule. The polymer ferrule will not deform stainless steel seats and can be reused. Except for the fitting for Rheodyne valves, these fittings are compatible with all 1/16 in. female compression-type fittings now used on HPLC columns.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)