Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

57691

RheFlex® PEEK Fittings

Short fitting set, 1/16 in. tubing

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
¥5,700

¥5,700


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 EA
¥5,700

About This Item

UNSPSC Code:
41115700

¥5,700


Check Cart for Availability

Request a Bulk Order

packaging

pkg of 5 ea

fitting

10-32 Rheodyne nuts and ferrules for 1/16 in. O.D. tubing

compatibility

1/16 in. tubing

Looking for similar products? Visit Product Comparison Guide

General description

PEEK (polyetheretherketone) fingertight fittings (short fitting set) are convenient, inert, and biocompatible. Use these fittings with 1/16 in. O.D. PEEK, stainless steel, titanium, Tefzel, or PTFE tubing. PEEK fittings are compatible with all HPLC solvents (avoid concentrated sulfuric and nitric acids), and can be used at temperatures up to 150° C. Unlike stainless steel ferrules, PEEK ferrules do not permanently lock into place on the tubing. This allows the interchanging of fittings with tubing and columns, from manufacturer to manufacturer (e.g. from Agilent to Waters), and still form a zero dead volume, leak-free connection.

Features and Benefits

For Rheodyne valves. Hold to 5000 psi (350 kg/cm²).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

RheFlex is a registered trademark of IDEX Health & Science, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service