Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

48392

Supelco

Triazine Pesticides Standard Mix

100 μg/mL each component in methanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

analytical standard

CofA

certificate of analysis is enclosed in each package.

feature

standard type calibration

packaging

ampule of 1 mL

concentration

100 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

multi-component solution

General description

Triazine derivatives are commonly used as herbicides, in order to control perennial weeds. They are very chemically stable and very persistent in environment. Triazine pesticides include atrazine, prometon, prometryn, propyzamide, terbutylazine and terbutryne.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Triazine Pesticides Standard Mix, was used as a standard, in double solid-phase extraction system, done in order to determine triazine herbicides in milk.

Other Notes

This product is now available as a certified reference solution. Please reference CRM48392.

Analyte

Description

    Ametryn
    Atrazine
    Prometon
    Prometryn
    Propazine
    Simazine
    Terbutryn

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

FSL

Group 4: Flammable liquids
Alcohols
Hazardous rank II

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

48392:4548173221052


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of double solid-phase extraction system for determining triazine herbicides in milk.
Lagana AM and Fago G.
Chromatographia, 41 (5-6), 175-182 (1995)
Multiresidue method for the simultaneous determination of four groups of pesticides in ground and drinking waters, using solid-phase microextraction-gas chromatography with electron-capture and thermionic specific detection.
Goncalves C and Alpendurada MF.
Journal of Chromatography A, 968 (1), 177-190 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service