Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

442298

Supelco

2,4,5,6-Tetrachloro-m-xylene

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6Cl4
CAS Number:
Molecular Weight:
243.95
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

room temp

SMILES string

Cc1c(Cl)c(C)c(Cl)c(Cl)c1Cl

InChI

1S/C8H6Cl4/c1-3-5(9)4(2)7(11)8(12)6(3)10/h1-2H3

InChI key

NTUBJKOTTSFEEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

442298:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ochan Otim
Environmental science. Processes & impacts, 21(9), 1580-1595 (2019-08-15)
Assessing the extent of ocean bed contamination by anthropogenic organic chemicals requires collecting and preserving the native state of sediments. The latter is particularly important since most sediment analyses are performed away from the sites of samples collection. Preservation, however
Xiang Wu et al.
Environmental pollution (Barking, Essex : 1987), 246, 124-130 (2018-12-12)
Predicting the translocation of organic contaminants to plants is crucial to ensure the quality of agricultural goods and assess the risk of human exposure through the food web. In this study, the performance of a modified plant uptake model was
Shanshan Yin et al.
Environment international, 135, 105402-105402 (2019-12-24)
Recent studies show that, even after being banned for agricultural applications for over 30 years, organochlorine pesticides (OCPs), including hexachlorocyclohexanes (HCHs) and dichlorodiphenyltrichloroethane (DDT) and its metabolites (DDXs), can still be found in various biological matrices and pose a potential hazard
Swatantra Pratap Singh et al.
The journal of physical chemistry. A, 121(27), 5156-5163 (2017-06-20)
The objective of this study was to elucidate the mechanism of abiotic hydrolysis of ES isomers, i.e., Endosulfan-1 (ES-1) and Endosulfan-2 (ES-2), using a combination of experiments and density functional theory (DFT) calculations. Hydrolysis of both ES-1 and ES-2 resulted
Shanshan Yin et al.
Environment international, 130, 104939-104939 (2019-06-30)
Currently, there is limited information about the mechanism of the human transplacental transfer for organochlorine pesticides (OCPs). This study aimed to evaluate the transplacental transfer of OCPs to better understand the influencing factors of exposure and transplacental efficiency. The study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service