Documents
24867
Supelco
Tube Holder for PAS-500 Pump
for use with 6mm sorbent tube
About This Item
UNSPSC Code:
41104008
technique(s)
active air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
compatibility
for use with 6mm sorbent tube
General description
Tube Holder for PAS-500 Pump for 6mm sorbent tubes is used as accessories for PAS-500 micro air sampler used in air monitoring.
Choose from one of the most recent versions:
Certificates of Analysis (COA)