Skip to Content
Merck
All Photos(1)

Key Documents

21013-U

Supelco

GC Stationary Phase

phase Bentone 34, bottle of 50 g

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
23151817
NACRES:
SB.54

form

solid

packaging

bottle of 50 g

parameter

0-180 °C temp. range

technique(s)

gas chromatography (GC): suitable

matrix active group

Bentone 34 phase

solubility

toluene: soluble

column type

packed GC

General description

GC methods are divided into two classes depending on the nature of stationary phases; gas-solid chromatography (GSC) and gas-liquid chromatography (GLC). GSC has solid adsorptive material and solute particles are removed from mobile phase by electrostatic forces. GLC has a thin layer of liquid coated or bonded on the surface of an inert particle or on the walls of the column where solute particles are retained in the liquid phase based on their partition coefficients.[1] The primary necessity of a stationary phase is to provide sample separation sustaining phase integrity over a reasonable period of time. It should be stable for the chemical and thermal changes. Selectivity, peak symmetry, analysis time, degree of separation, peak tailing are few parameters that should be considered before choosing a stationary phase.[2] Bentone 34 is a bentonite treated with dimethyloctadecylammonium salts which has broad applications in GC. It can differentiate between isomers of many aromatic compounds and cycloalkane derivatives. It can also be modified with silicone oil in order to improve the peak symmetry.[3]
Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase.

Application

Bentone 34 may be used as column coating, using a benzene-methanol (9:1) mixture as dispersion agent to analyse cresol mixtures.[4]

Kit Components Only

Product No.
Description

  • Bentone® 34

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Rotzsche
Stationary Phases in Gas Chromatography (1991)
Analysis of isomeric cresols by gas chromatography.
Nardillo, A. M.,
Chromatographia, 32, 457-460 (1991)
David B. Troy, Paul Beringer
Remington: The Science and Practice of Pharmacy, 605-605 (2006)
James P. Lodge, Jr.
Methods of Air Sampling and Analysis, 98-98 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service