Safety Information
20857
Tubing Bender
Three-Size for 1/4in. (6 mm, 3/16in. (4 mm) & 1/8in. tube
About This Item
UNSPSC Code:
41100000
General description
The heavy duty benders easily bend aluminum, copper, or stainless steel tubing up to 180°, without kinking or splitting the tubing.
The three-in-one tool (20857) bends 1/8 in., 3/16 in., or 1/4 in. (3, 4, or 6 mm) tubing of standard wall thickness.
20422-U has a bend radius of 3/8 inch from center.
20424-U has a bend radius of 9/16 inch from center.
20857 (three-in-one tool) has a bend radius of 14.2 mm from center (pictured above).
Choose from one of the most recent versions:
Certificates of Analysis (COA)