Skip to Content
Merck
All Photos(1)

Documents

Safety Information

18927

Supelco

2-[2-[[3-[2-(Dimethylamino)ethyl]-1H-indol-5-yl]methyl]-5-(1H-1,2,4-triazol-1-ylmethyl)-1H-indol-3-yl]-N,N-dimethylethanamine

reference material

Synonym(s):

Rizatriptan Benzoate impurity A (PhEur)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H35N7
CAS Number:
Molecular Weight:
469.62
Beilstein:
57609764

grade

reference material

API family

rizatriptan

Assay

≥90.0% (HPLC)

form

solid

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

SMILES string

CN(C)CCC1=CNC(C1=C2)=CC=C2CC3=C(CCN(C)C)C4=CC(CN5N=CN=C5)=CC=C4N3

InChI

1S/C28H35N7/c1-33(2)11-9-22-16-30-26-7-5-20(13-24(22)26)15-28-23(10-12-34(3)4)25-14-21(6-8-27(25)32-28)17-35-19-29-18-31-35/h5-8,13-14,16,18-19,30,32H,9-12,15,17H2,1-4H3

InChI key

WMKBUQPKAAJBHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

18927-100MG:
18927-VAR:
18927-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service