Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

13184-U

Supelco

Dowex 21K XLT Resin 100 grams

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115710
NACRES:
SB.52

form

beads

crosslinking

<10 % cross-linked

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene

matrix active group

quaternary amine

particle size

525-625 μm

capacity

1.4 meq/mL

separation technique

anion exchange

General description

Dowex 21K XLT is a strong basic anionic exchange resin, selectively adsorbs gold-cyanide complex. Dowex is used as a solid acid catalyst in organic synthesis. Dowex polymer is highly efficient solid acid catalyst for the regioselective ring opening of epoxides to β-hydroxy thiocyanates.

Application

Dowex 21K XLT was used:
  • to detect the existence of gold-cyanide complex using Scanning electron microscopy and Energy dispersive X-ray spectroscopy
  • to remove excess amount of arsenate during the analysis of arsines using atomic fluorescence spectrometer.

Legal Information

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

13184-U:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dowex as Reusable Acidic Polymeric Catalyst in the Efficient and Regioselective Conversion of Epoxides into β-Hydroxy Thiocyanates under Solvent Free Conditions.
Kiasat, Ali Reza, and Mehdi Fallah-Mehrjardi.
J. Chin. Chem. Soc., 55, 1119-1124 (2008)
Selective adsorption of the gold-cyanide complex from waste rinse water using Dowex 21K XLT resin
Ok, Yong Sik, and Choong Jeon.
Journal of Industrial and Engineering Chemistry (Amsterdam, Netherlands), 20, 1308-1312 (2014)
Efrén Ordóñez et al.
The Journal of biological chemistry, 284(22), 15107-15116 (2009-03-17)
We identified the first enzymes that use mycothiol and mycoredoxin in a thiol/disulfide redox cascade. The enzymes are two arsenate reductases from Corynebacterium glutamicum (Cg_ArsC1 and Cg_ArsC2), which play a key role in the defense against arsenate. In vivo knockouts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service