Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

06768

Supelco

Buturon

reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13ClN2O
CAS Number:
Molecular Weight:
236.70
Beilstein:
2941811
MDL number:
UNSPSC Code:
41116107

grade

reference material

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

−20°C

SMILES string

CC(C#C)N(C)C(=O)Nc1ccc(Cl)cc1

InChI

1S/C12H13ClN2O/c1-4-9(2)15(3)12(16)14-11-7-5-10(13)6-8-11/h1,5-9H,2-3H3,(H,14,16)

InChI key

BYYMILHAKOURNM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Buturon belongs to the class of phenyl-urea herbicides, widely used in agriculture since they can inhibit the process of photosynthesis in weeds. These herbicides are employed for the selective or nonselective control of many annual and perennial weeds in corn, soybean, cotton, rice, and potato crops.
This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Application

Buturon may be used as a reference material for the quantification of the analyte in aqueous samples, rice, corn, food, and environmental samples using chromatography techniques.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

06768-VAR:
06768-BULK:
06768-50MG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous testing of multiclass organic contaminants in food and environment by liquid chromatography/dielectric barrier discharge ionization-mass spectrometry
Gilbert-Lopez B, et al.
Analyst, 137(22), 5403-5410 (2012)
Simultaneous determination of 15 phenylurea herbicides in rice and corn using HPLC with fluorescence detection combined with UV decomposition and post-column derivatization.
Mou R-X, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 875(2), 437-443 (2008)
Solid-phase microextraction coupled with high-performance liquid chromatography for the determination of phenylurea herbicides in aqueous samples.
Lin HH, et al.
Journal of Chromatography A, 1012(1), 57-66 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service