Skip to Content
Merck
All Photos(1)

Key Documents

Z722464

IKA® ULTRA-TURRAX® dispersers

T-25 Digital, for volumes of 1-2,000 mL, 230 V, 1/cs

Synonym(s):

tissue grinders, tissue homogenizers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103800
NACRES:
NB.73
Product Z722464 is not currently sold in your country. Contact Technical Service

AC/DC input

230 V AC

description

sample 1 - 2,000 mL

feature

CE compliant

packaging

pack of 1 ea

manufacturer/tradename

IKA 3725000

parameter

3000-25000 rpm speed

Looking for similar products? Visit Product Comparison Guide

General description

T 25 Digital ULTRA-TURRAX®

  • The spectrum of applications ranges from homogenizing waste water samples to the use in laboratory reactors, to dispersion tasks under vacuum/pressure and sample preparation in medical diagnostics
  • For volumes of 1-2,000 mL (H20)
  • Three types of shaft bearings (sold separately)
  • Standard version with digital display supplied only
  • Rotor-Stator configurations have thirty years of proven, guaranteed comparability of test results

Application

  • Particle reduction, emulsifications, and reaction enhancements in chemical, pharmaceutical, and food research
  • Tissue homogenization for biological and medical research
  • Alcohol production
  • Heavy metals analysis in sludge samples
  • Homogenization of marine samples for separating bacteria aggregates

Note: The dispersing element/rod is sold separately

Legal Information

IKA is a registered trademark of IKA-Werke GmbH & Co. KG
ULTRA-TURRAX is a registered trademark of IKA-Werke GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Can we use it for Sweet Condensed Milk with fat content 10%, sugar 50% and milk solids 25%?

    1 answer
    1. This is a product of IKA and is cited for use in homogenizing tissues and samples from various sources. The maximum rated viscosity is 5000 mPas. While Sweet Condensed Milk is not specifically mentioned, the viscosity is within the maximum limits.

      Note that this product is for research use only. Unless otherwise stated, this product is intended for research use only and not to be used for any other purpose, which includes but is not limited to unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses, or any type of consumption or application to humans or animals.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service