Key Documents
Z719943
Sigma-Aldrich
Microcentrifuge Tube Opener
double-ended tool opens both small and larger tubes
About This Item
UNSPSC Code:
41121702
NACRES:
NB.11
manufacturer/tradename
Bel-Art
technique(s)
cryopreservation: suitable
General description
Easily open microcentrifuge tubes with this double-ended tool. Works for all sizes of tubes. One end opens standard 1.5, 2 and 2.2 mL tubes while the other end has a forked blade that opens smaller sizes (250, 400 and 550 μL tubes). 79mm (31/8 in.) long.
Choose from one of the most recent versions:
Certificates of Analysis (COA)