Key Documents
Z379417
Sigma-Aldrich
Hanna Piccolo® pH meter
Piccolo 2
About This Item
UNSPSC Code:
41115603
description
temperature compensation 0 - 70 °C (automatic with integral temperature sensor)
feature
calibration manual dual point, ±1 pH
manufacturer/tradename
HI98112
pH measuring range
0-14, accuracy: 0.01, resolution: 0.01
electrode L
160 mm
weight
400 gincluding calibration screwdriver, ABS carrying case, and batteries
70 g
battery power
3 × 1.4 V button cell, 100 hr life (continuous operation)
General description
Hand-held stick-style pH/temperature meters with rugged design for field use. "Amplified electrode" delivers strong signal to the meter, eliminating the vulnerability to noise, humidity, and dirt that is characteristic of conventional meters.
Legal Information
Piccolo is a registered trademark of Hanna Instruments
Choose from one of the most recent versions:
Certificates of Analysis (COA)