Skip to Content
Merck
All Photos(2)

Key Documents

W4769

Sigma-Aldrich

Monoclonal Anti-WIPI-1 antibody produced in mouse

~1.5 mg/mL, clone WIPI1-4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ATG18, Anti-WIPI-1α, Anti-WIPI49

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

WIPI1-4, monoclonal

form

buffered aqueous solution

mol wt

antigen ~49 kDa

species reactivity

human, mouse, bovine, rat

concentration

~1.5 mg/mL

technique(s)

western blot: 2-4 μg/mL using whole extract of G361 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WIPI1(55062)
mouse ... Wipi1(52639)
rat ... Wipi1(303630)

General description

Monoclonal Anti-WIPI-1 (mouse IgG1 isotype) is derived from the hybridoma WIPI1-4 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to a fragment of human WIPI-1, conjugated to KLH. WIPI-1 (WD repeat domain phosphoinositide-interacting protein 1) the mammalian orthologue of Atg18 in S. cerevisiae and A. thaliana, is a member of the WIPI subfamily of WD-repeat (Trp-Asp) proteins. WIPI-1 has a 7-bladed propeller structure and contains a conserved motif for interaction with phospholipids. WIPI-1 is expressed in normal human tissues with high levels in skeletal muscle, heart and testis.

Application

Monoclonal Anti-WIPI-1 antibody has been used in immunoblotting, immunoprecipitation.

Biochem/physiol Actions

WIPI-1 (WD repeat domain phosphoinositide-interacting protein 1) plays an important role in signal transduction, transcription regulation and apoptosis. Endogenous WIPI-1 partially colocalizes with the autophagosomal marker LC3 and induction of autophagy leads to the accumulation of WIPI-1 in large vesicular and cup-shaped structures that are characteristic for autophagy-linked proteins. WIPI-1 is aberrantly expressed in human cancer and is linked to starvation induced autophagy.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human WIPI-1 puncta-formation: a novel assay to assess mammalian autophagy
Proikas-Cezanne T, et al.
Febs Letters, 581(18), 3396-3404 (2007)
WIPI-1alpha(WIPI49), a member of the novel 7-bladed WIPI protein family, is aberrantly expressed in human cancer and is linked to starvation-induced autophagy
Proikas-Cezanne T, et al.
Oncogene, 23(58), 9314-9314 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service