Skip to Content
Merck
All Photos(1)

Documents

TMO374502

Nunc® latch rack

polypropylene, capacity 1.8 mL, flat bottom natural, sterile, case of 480 ea

Synonym(s):

racked universal tube, sample storage tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122804
NACRES:
NB.11

material

flat bottom natural
polypropylene

sterility

sterile

packaging

case of 480 ea

manufacturer/tradename

Nunc 374502

technique(s)

PCR: suitable

capacity

1.8 mL

General description

“Nunc” is an automation friendly 1.8 mL externally threaded universal tube to enhance the efficacy and throughput of your sample storage workflow. The latch rack features an ANSI SBS footprint that is compatible with automation platforms for liquid handling and sample storage. These tubes are available in racked or bulk formats, with options for 2D barcodes or non-coded tubes. Additionally, empty racks are also available for your convenience.

Features and Benefits

Scalable.
  • Tubes and Racks: Ideal for both lower-throughput and high-throughput use.
  • Feature graduations and white patch for manual marking.
  • Compatible with traditional cryostorage boxes designed for 10x10 arrays.
  • External threads limit contamination.
  • Automation-pickable in Universal Latch Rack.


Innovative Universal Cap Design:
  • Features a single-piece construction with a molded-in gasket.
  • Designed to prevent over-tightening, both manually and with automated capping tools.
  • The gasket material is made from thermoplastic elastomer, ensuring compatibility with storage at room temperature and down to the vapor phase of liquid nitrogen.


Secure Tracking:
  • The base of each tube features a permanently bonded, unique 2D barcode laser-etched for secure sample identification and tracking.
  • Instant scanning by 2D barcode readers enables easy registration in any application or database, with various export options available.
  • Access sample information and identify contents by simply scanning the tube.
  • Tube 374502NOV has a 1D (linear) barcode and a human-readable code on the side, corresponding to the 2D barcode on the bottom of the tube.


Stringent Quality Control:
  • Scanning is performed on each 2D barcoded storage tube to ensure readability.
  • The barcodes undergo cross-checking with a database of previously assigned codes to avoid duplicates.
  • Every tube is manufactured with class VI medical-grade resins and has a sterility assurance level of 10^6.

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service