Skip to Content
Merck
All Photos(1)

Documents

SYNT100024

Synthware NMR tube washer

borosilicate glass 3.3, joint: ST/NS 24/40

Synonym(s):

Synthware T100024

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

material

borosilicate glass 3.3

Joint

joint: ST/NS24/40

packaging

pack of 1 ea

manufacturer/tradename

Kemtech T100024

General description

Nmr Tube Washer, 24/40

Application

kemetch; synthware; nmr; nmr washer

Features and Benefits

Organic Chemistry

Legal Information

Synthware is a trademark of Kemtech America Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service