SYNS380216
Synthware™ long syringe needle for solvent retrieval
Gauge 18, O.D. × L 1.2 mm × 180 mm , Bevel Tip, stainless steel
Synonym(s):
Synthware S380216, stainless steel syringe needle, bevel tip needle
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Kemtech S380216
O.D. × L
1.2 mm × 180 mm , Bevel Tip
needle gauge
18 , Bevel Tip
Looking for similar products? Visit Product Comparison Guide
General description
Syringe Needle, Stainless Steel, 1.2mm, 18G, 180mm
Application
kemetch; synthware; syrnge needle; stainless steel
Features and Benefits
Organic Chemistry
Legal Information
Synthware is a trademark of Kemtech America Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service