SYNB135421
Synthware™ vacuum desiccator
includes a ceramic plate and lid with stopcock/port, ID 210mm
Synonym(s):
Synthware B135421, desiccator with Top Knob
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Kemtech B135421
diam.
210 mm
Looking for similar products? Visit Product Comparison Guide
General description
Vacuum Desiccator, ID 210mm, Includes Lid & Ceramic Plate includes Accessories: A682440G
Application
kemetch; synthware; vacuum desiccator; desiccator
Features and Benefits
Organic Chemistry
Legal Information
Synthware is a trademark of Kemtech America Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service