Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

SML3204

Sigma-Aldrich

Dapansutrile

≥98% (HPLC)

Synonym(s):

3-(Methylsulfonyl)propanenitrile; 3-(Methylsulfonyl)propionitrile, Dapansutrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NO2S
CAS Number:
Molecular Weight:
133.17
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=S(C)(CCC#N)=O

InChI key

LQFRYKBDZNPJSW-UHFFFAOYSA-N

Biochem/physiol Actions

Dapansutrile is an orally active, specific and potent inhibitor of NLRP3 inflammasome activation that decreases IL-1β and IL-18 release. Dapansutrile exhibits no effect on NLRC4 and AIM2 inflammasomes activation. It appears that Dapansutrile directly interact with NLRP3. It does not show hematological, or organ toxicities in humans at oral doses up to 1,000 mg/day
orally active, specific and potent inhibitor of NLRP3 inflammasome activation; anti-inflammatory

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Deleterious substance

JAN Code

SML3204-VAR:
SML3204-25MG:
SML3204-5MG:
SML3204-BULK:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The NLRP3 Inflammasome Inhibitor, OLT1177 (Dapansutrile), Reduces Infarct Size and Preserves Contractile Function After Ischemia Reperfusion Injury in the Mouse
Journal of Cardiovascular Pharmacology, 73(4), 215-222 (2019)
OLT1177, a ?-sulfonyl nitrile compound, safe in humans, inhibits the NLRP3 inflammasome and reverses the metabolic cost of inflammation
Proceedings of the National Academy of Sciences of the USA, 115(7), E1530-E1539 (2018)
NLRP3 inflammasome inhibitor OLT1177 suppresses joint inflammation in murine models of acute arthritis
Arthritis Research & Therapy, 20(1), 169-169 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service