Skip to Content
Merck
All Photos(1)

Documents

SML1863

Sigma-Aldrich

NSC57969

≥98% (HPLC)

Synonym(s):

7-(1-Piperidinylmethyl)-8-quinolinol, 7-(Piperidinomethyl)-8-quinolinol, 8-Hydroxy-7-(piperidinomethyl)quinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18N2O
CAS Number:
Molecular Weight:
242.32
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

OC1=C(N=CC=C2)C2=CC=C1CN3CCCCC3

InChI

1S/C15H18N2O/c18-15-13(11-17-9-2-1-3-10-17)7-6-12-5-4-8-16-14(12)15/h4-8,18H,1-3,9-11H2

InChI key

WXMFPLZGDXLEMN-UHFFFAOYSA-N

Biochem/physiol Actions

NSC57969 is a MDR-selective compound that exhibits a robust Pgp-dependent toxic activity across diverse cancer cell lines. NSC57969 shows profound toxicity against doxorubicin resistant Brca1-/-;p53-/- spontaneous mouse mammary carcinoma cells. NSC57969 eradicates P-glycoprotein expression in doxorubicin resistant brca1-/-;p53-/- spontaneous mouse mammary carcinoma, and MES-SA/Dx5 cells. However treatment of normal cells (hCMEC/D3 cells) with NSC57969 does not induce loss of Pgp.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

András Füredi et al.
Molecular cancer therapeutics, 16(1), 45-56 (2016-10-21)
Despite significant progress, resistance to chemotherapy is still the main reason why cancer remains a deadly disease. An attractive strategy is to target the collateral sensitivity of otherwise multidrug resistant (MDR) cancer. In this study, our aim was to catalog

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service