Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

SE120006

Sigma-Aldrich

Total Aflatoxin ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

technique(s)

ELISA: suitable
competitive inhibition ELISA: suitable

assay range

inter-assay cv: <15%
intra-assay cv: <10%
sensitivity: 0-4 ng/mL
standard curve range: 0.2-4 ng/mL

shipped in

wet ice

storage temp.

2-8°C

General description

The Sigma-Aldrich® Total Aflatoxin ELISA (Enzyme-Linked Immunosorbent Assay) kit is a solid phase direct competitive enzyme immunoassay. An aflatoxin specific antibody optimized to cross react with all four subtypes of aflatoxin (see cross reactivity information) is coated to a polystyrene microwell. Toxins are extracted from a ground sample with 70% methanol. The extracted sample and HRP-conjugated aflatoxin B1 are mixed and added to the antibody-coated microwell. Aflatoxin from the extracted sample and HRP-conjugated aflatoxin B1 compete to bind with the antibody coated to the microwell. Microwell contents are decanted and non-specific reactants are removed by washing. An enzyme substrate (TMB) is added and colour (blue) develops. The intensity of the colour is directly proportional to the amount of bound conjugate and inversely proportional to the concentration of aflatoxin in the sample or standard. Therefore, as the concentration of aflatoxin in the sample or standard increases, the intensity of the blue colour will decrease.

Legal Information

SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 1B - Ox. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 1

Storage Class Code

5.1B - Oxidizing hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Please refer to KIT Component information

PRTR

Please refer to KIT Component information

FSL

Please refer to KIT Component information

ISHL Indicated Name

Please refer to KIT Component information

ISHL Notified Names

Please refer to KIT Component information

Cartagena Act

Please refer to KIT Component information

JAN Code

キットコンポーネントの情報を参照してください


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service