Skip to Content
Merck
All Photos(1)

Key Documents

SCP0218

Sigma-Aldrich

PPlases Substrate

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H41N6O9
Molecular Weight:
665.71
UNSPSC Code:
12352204
NACRES:
NA.32
Product SCP0218 is not currently sold in your country. Contact Technical Service

Product Name

PPlases Substrate, calorimetric,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥80%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Suc-Ala-Leu-Pro-Phe-pNA

Application

Suc-Ala-Leu-Pro-Phe-pNA, colorimetric and Suc-Ala-Leu-Pro-Phe-AMC, fluorogenic are used as substrates for peptidyl-prolyl cis/trans isomerases (PPIases, EC: 5.2.1.8).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service