Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

SAB5702064

Sigma-Aldrich

Anti-HGD Antibody, clone 7O8Q1, Rabbit Monoclonal

Synonym(s):

HGD, AKU, HGO, homogentisate 1,2-dioxygenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

7O8Q1, monoclonal

form

liquid

mol wt

50 kDa

species reactivity

human

concentration

0.4 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

IRGHYEAKQGGFLPGGGSLHSTMTPHGPDADCFEKASKVKLAPERIADGTMAFMFESSLSLAVTKWGLKASRCLDENYHKCWEPLKSHFTPNSRNPAEPN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HGD(3081)

General description

This gene encodes the enzyme homogentisate 1,2 dioxygenase. This enzyme is involved in the catabolism of the amino acids tyrosine and phenylalanine. Mutations in this gene are the cause of the autosomal recessive metabolism disorder alkaptonuria.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 346-445 of human HGD (Q93099).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

SAB5702064-100UL:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service