Skip to Content
Merck
All Photos(2)

Key Documents

Safety Information

SAB1409916

Sigma-Aldrich

Anti-AP3B1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

ADTB3, ADTB3A, HPS, HPS2, PE

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
¥60,750

¥60,750


Estimated to ship onApril 25, 2025



Select a Size

Change View
50 μG
¥60,750

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

¥60,750


Estimated to ship onApril 25, 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 121.3 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AP3B1(8546)

Related Categories

General description

This gene encodes a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. The encoded protein is part of the heterotetrameric AP-3 protein complex which interacts with the scaffolding protein clathrin. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 2. (provided by RefSeq)

Immunogen

AP3B1 (NP_003655.3, 1 a.a. ~ 1094 a.a) full-length human protein.

Sequence
MSSNSFPYNEQSGGGEATELGQEATSTISPSGAFGLFSSDLKKNEDLKQMLESNKDSAKLDAMKRIVGMIAKGKNASELFPAVVKNVASKNIEIKKLVYVYLVRYAEEQQDLALLSISTFQRALKDPNQLIRASALRVLSSIRVPIIVPIMMLAIKEASADLSPYVRKNAAHAIQKLYSLDPEQKEMLIEVIEKLLKDKSTLVAGSVVMAFEEVCPDRIDLIHKNYRKLCNLLVDVEEWGQVVIIHMLTRYARTQFVSPWKEGDELEDNGKNFYESDDDQKEKTDKKKKPYTMDPDHRLLIRNTKPLLQSRNAAVVMAVAQLYWHISPKSEAGIISKSLVRLLRSNREVQYIVLQNIATMSIQRKGMFEPYLKSFYVRSTDPTMIKTLKLEILTNLANEANISTLLREFQTYVKSQDKQFAAATIQTIGRCATNILEVTDTCLNGLVCLLSNRDEIVVAESVVVIKKLLQMQPAQHGEIIKHMAKLLDSITVPVARASILWLIGENCERVPKIAPDVLRKMAKSFTSEDDLVKLQILNLGAKLYLTNSKQTKLLTQYILNLGKYDQNYDIRDRTRFIRQLIVPNVKSGALSKYAKKIFLAQKPAPLLESPFKDRDHFQLGTLSHTLNIKATGYLELSNWPEVAPDPSVRNVEVIELAKEWTPAGKAKQENSAKKFYSESEEEEDSSDSSSDSESESGSESGEQGESGEEGDSNEDSSEDSSSEQDSESGRESGLENKRTAKRNSKAKGKSDSEDGEKENEKSKTSDSSNDESSSIEDSSSDSESESEPESESESRRVTKEKEKKTKQDRTPLTKDVSLLDLDDFNPVSTPVALPTPALSPSLMADLEGLHLSTSSSVISVSTPAFVPTKTHVLLHRMSGKGLAAHYFFPRQPCIFGDKMVSIQITLNNTTDRKIENIHIGEKKLPIGMKMHVFNPIDSLEPEGSITVSMGIDFCDSTQTASFQLCTKDDCFNVNIQPPVGELLLPVAMSEKDFKKEQGVLTGMNETSAVIIAAPQNFTPSVIFQKVVNVANVGAVPSGQDNIHRFAAKTVHSGSLMLVTVELKEGSTAQLIINTEKTVIGSVLLRELKPVLSQG

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

JAN Code

SAB1409916-50UG:


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service