Skip to Content
Merck
All Photos(1)

Documents

Safety Information

S5525

Sigma-Aldrich

SigmaClean water bath treatment

bottle of 4 oz

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41103706
NACRES:
NB.77

description

not available in EU; EU alternative Z611662

packaging

bottle of 4 oz

availability

not available in EU (EU customers: alternative Z611662)

InChI

1S/C23H42N.ClH/c1-4-5-6-7-8-9-10-11-12-13-14-18-21-24(2,3)22-23-19-16-15-17-20-23;/h15-17,19-20H,4-14,18,21-22H2,1-3H3;1H/q+1;/p-1

InChI key

OCBHHZMJRVXXQK-UHFFFAOYSA-M

General description

The 4 oz (or ~118.3 mL) bottle of SigmaClean water bath treatment can be used to treat about 8 gallons (or 30.2 L) of water.

These instructions provide an extremely concentrated bath solution. S5525 is still effective even at 0.0025 oz of the treatment to 1 gallon of water.

Please note: If you go below 0.0025 oz, the treatment will be too dilute to be effective.

Legal Information

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

129.0 °F

Flash Point(C)

53.9 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PRTR

Class I Designated Chemical Substances

FSL

Group 4: Flammable liquids
Type 2 petroleums
Hazardous rank III
Water insoluble liquid

ISHL Indicated Name

Substances Subject to be Indicated Names

ISHL Notified Names

Substances Subject to be Notified Names

JAN Code

S5525-4OZ:4548173951355
S5525-4OZPW:
S5525-BULK:


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service