Skip to Content
Merck
All Photos(1)

Documents

Safety Information

RAB1082

Sigma-Aldrich

Human Serpin G1 ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum sample(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2.4 ng/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SERPING1(710)

General description

This ELISA antibody pair detects human SERPING1. Other species not yet determined.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Please refer to KIT Component information

PRTR

Please refer to KIT Component information

FSL

Please refer to KIT Component information

ISHL Indicated Name

Please refer to KIT Component information

ISHL Notified Names

Please refer to KIT Component information

Cartagena Act

Please refer to KIT Component information

JAN Code

キットコンポーネントの情報を参照してください


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service