Skip to Content
Merck
All Photos(1)

Key Documents

Safety Information

RAB0355

Sigma-Aldrich

Phospho-Met (pTyr1234) ELISA Kit

for detection of phospho-met (pTyr1234) in human or mouse cell and tissue lysates

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
¥109,000

¥109,000


Estimated to ship onApril 16, 2025

New, lower price on this item!


Select a Size

Change View
1 KIT
¥109,000

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

¥109,000


Estimated to ship onApril 16, 2025

New, lower price on this item!

species reactivity

human, mouse

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell lysate
sample type tissue lysate

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MET(4233)

General description

The Phospho-Met (pTyr1234) ELISA (Enzyme-Linked Immunosorbent Assay) kit is a very rapid, convenient and sensitive assay kit that can monitor the activation or function of important biological pathways in cell lysates.

Immunogen

Met (pTyr1234) synthetic phosphopeptide

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Met. Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

PDSCL

Please refer to KIT Component information

PRTR

Please refer to KIT Component information

FSL

Please refer to KIT Component information

ISHL Indicated Name

Please refer to KIT Component information

ISHL Notified Names

Please refer to KIT Component information

Cartagena Act

Please refer to KIT Component information

JAN Code

キットコンポーネントの情報を参照してください


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service